Learn More
Mettler Toledo™ InLab™ 605 ISM Intelligent Dissolved Oxygen Sensors
Product Code. 10379604 Shop All Mettler Toledo Products
Change view
Click to view available options
Length (Metric) Cable:
10 m
5 m
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Cable unitSize
10379604 5 m Each
10126064 10 m Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10379604 Supplier Mettler Toledo™ Supplier No. 51344612
Specifications
Length (Metric) Cable 5 m
Temperature (Metric) Range 0°C to 60°C
Certifications/Compliance CE, ISO 9001 certified