Learn More
Miele Professional™ Injector Nozzle
Product Code. 10764523 Shop All Miele Products
Change view
Click to view available options
Model:
ID 110
ID 140
ID 160
ID 180
ID 240
ID 90
Dimensions (Dia. x H):
2.5 dia. x 110mm L
2.5 dia. x 90mm L
4 dia. x 140mm L
4 dia. x 160mm L
4 dia. x 180mm L
6 dia. x 240 mm L
Unit Size:
Each
Product Code. Model Dimensions (Dia. x H) unitSize
10764523 ID 140 4 dia. x 140mm L Each
10486402 ID 90 2.5 dia. x 90mm L Each
10008441 ID 110 2.5 dia. x 110mm L Each
10712993 ID 160 4 dia. x 160mm L Each
10231572 ID 180 4 dia. x 180mm L Each
10640384 ID 240 6 dia. x 240 mm L Each
6 options
Product Code. 10764523
Brand: Miele Professional™ 3810340
Only null left
Add to basket
Add to basket
Injector nozzles with plastic supports
  • Suitable for standard mobile injector units and injector baskets
  • Suitable for measuring flasks and test tubes, for example
TRUSTED_SUSTAINABILITY
Specifications
Material Stainless Steel
Product Type Injector Nozzle
Color Stainless Steel
Model ID 140
For Use With (Equipment) Washer-disinfectors, laboratory
Includes Plastic support
Dimensions (Dia. x H) 4 dia. x 140mm L
Quantity 1 Each