Learn More
Miele Professional™ Injector nozzle
Product Code. 10454635 Shop All Miele Products
Change view
Click to view available options
:
Depth: 220 mm
Product Code.
10454635 Depth: 220 mm
1 options
Product Code. 10454635
Brand: Miele Professional™ 3809540
This item requires a price update prior to being ordered.
Request a Price for this item here.
Clamps E 354, height-adjustable for optimum positioning of laboratory glassware on the nozzle
  • Suitable for injector nozzle E 352
  • Height adjustable locking
  • Suitable for bottles, Erlenmeyer flasks and round bottomed flasks for example
TRUSTED_SUSTAINABILITY
Specifications
Description Clamps
Material Stainless Steel
Color Stainless Steel
For Use With (Equipment) Labwasher
Depth (Metric) 220 mm