missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Inhibin beta A Mouse anti-Human, Porcine, Clone: 1F8D8, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 60352-1-IG-150UL
This item is not returnable.
View return policy
Description
Activins are dimeric growth and differentiation factors which belong to the transforming growth factor-beta (TGF-beta) superfamily of structurally related signaling proteins. Activins signal through a heteromeric complex of receptor serine kinases which include at least two type I ( I and IB) and two type II (II and IIB) receptors. These receptors are all transmembrane proteins, composed of a ligand-binding extracellular domain with cysteine-rich region, a transmembrane domain, and a cytoplasmic domain with predicted serine/threonine specificity. Type I receptors are essential for signaling; and type II receptors are required for binding ligands and for expression of type I receptors. Type I and II receptors form a stable complex after ligand binding, resulting in phosphorylation of type I receptors by type II receptors. Activin A mutations are associated with fibrodysplasia ossificans progressive.Specifications
| Inhibin beta A | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P03970, P08476 | |
| INHBA | |
| Peptide | |
| 150 μL | |
| Primary | |
| Human, Pig | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1F8D8 | |
| Unconjugated | |
| INHBA | |
| Activin beta A chain, EDF, FRP, INHBA, INHBA-Specific, Inhibin beta A chain, inhibin, beta A | |
| Mouse | |
| Protein G | |
| RUO | |
| 3624, 397093 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction