missing translation for 'onlineSavingsMsg'
Learn More

Inhibin beta A Mouse anti-Human, Porcine, Clone: 1F8D8, Proteintech

Product Code. p-7213004 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16847475 20 μL 20µL
16837475 150 μL 150µL
2 options
This item is not returnable. View return policy

Product Code. 16847475

Brand: Proteintech 603521IG20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Activins are dimeric growth and differentiation factors which belong to the transforming growth factor-beta (TGF-beta) superfamily of structurally related signaling proteins. Activins signal through a heteromeric complex of receptor serine kinases which include at least two type I ( I and IB) and two type II (II and IIB) receptors. These receptors are all transmembrane proteins, composed of a ligand-binding extracellular domain with cysteine-rich region, a transmembrane domain, and a cytoplasmic domain with predicted serine/threonine specificity. Type I receptors are essential for signaling; and type II receptors are required for binding ligands and for expression of type I receptors. Type I and II receptors form a stable complex after ligand binding, resulting in phosphorylation of type I receptors by type II receptors. Activin A mutations are associated with fibrodysplasia ossificans progressive.

Specifications

Antigen Inhibin beta A
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 1F8D8
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene INHBA
Gene Accession No. P03970, P08476
Gene Alias Activin beta A chain, EDF, FRP, INHBA, INHBA-Specific, Inhibin beta A chain, inhibin, beta A
Gene Symbols INHBA
Host Species Mouse
Immunogen Peptide
Purification Method Protein G
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3624, 397093
Target Species Human, Pig
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.