Learn More
Fisherbrand™ Individual Stainless Steel Cylindrical Calibration Weights, Class M1 with certificate
Product Code. 15575159 Shop All Fisherbrand Products
Change view
Click to view available options
Weight (Metric):
1 g
2 g
5 g
10 g
20 g
50 g
100 g
200 g
500 g
1 kg
2 kg
5 kg
10 kg
Unit Size:
1 unit
Product Code. Weight (Metric) unitSize
15575159 50 g 1 unit
15535179 1 kg 1 unit
15535159 20 g 1 unit
15575139 1 g 1 unit
15575179 2 kg 1 unit
15595149 10 g 1 unit
15595169 500 g 1 unit
15595189 10 kg 1 unit
15555149 5 g 1 unit
15555169 200 g 1 unit
15515189 5 kg 1 unit
15515169 100 g 1 unit
15515149 2 g 1 unit
13 options
Product Code. 15575159
Only null left
Add to basket
Add to basket
Use with weighing instruments of medium accuracy class III. Fisherbrand™ Class M1 Stainless Steel Calibration Weights are ideal for the verification or calibration of class M2 weights.
  • Class M1 stainless steel calibration weight
  • Delivered with a calibration certificate guaranteeing traceability
  • In compliance with OIML R111
Specifications
Classification M1
Material Stainless Steel
Weight (Metric) 50 g
Mass 50 g
Shape Cylindrical