Learn More
Fisherbrand™ Individual Stainless Steel Cylindrical Calibration Weights, Class F2 with certificate
Product Code. 15575169 Shop All Fisherbrand Products
Change view
Click to view available options
Weight (Metric):
1 g
2 g
5 g
10 g
20 g
50 g
100 g
200 g
500 g
1 kg
2 kg
5 kg
10 kg
Unit Size:
1 unit
Product Code. Weight (Metric) unitSize
15575169 500 g 1 unit
15535169 200 g 1 unit
15535149 5 g 1 unit
15575149 10 g 1 unit
15575189 10 kg 1 unit
15595139 2 g 1 unit
15595159 100 g 1 unit
15595179 5 kg 1 unit
15555139 1 g 1 unit
15555159 50 g 1 unit
15555179 2 kg 1 unit
15515179 1 kg 1 unit
15515159 20 g 1 unit
13 options
Product Code. 15575169
Only null left
Add to basket
Add to basket
Use with weighing instruments of high accuracy class II. Fisherbrand™ Class F2 Stainless Steel Calibration Weights are also intended for use in important commercial transactions, such as those involving precious metals and stones.
  • Class F2 stainless steel calibration weight
  • Weights intended for use in the verification or calibration of class M1 and possibly class M2 weights
  • Delivered with a calibration certificate guaranteeing traceability
  • In compliance with OIML R111
Specifications
Classification F2
Material Stainless Steel
Weight (Metric) 500 g
Mass 500 g
Shape Cylindrical