Learn More
Fisherbrand™ Individual Stainless Steel Cylindrical Calibration Weights, Class F1
Product Code. 15525059 Shop All Fisherbrand Products
Change view
Click to view available options
Weight (Metric):
1 g
2 g
5 g
10 g
20 g
50 g
100 g
200 g
500 g
1 kg
2 kg
5 kg
10 kg
Unit Size:
1 unit
Product Code. Weight (Metric) unitSize
15525059 100 g 1 unit
15565049 1 g 1 unit
15535059 200 g 1 unit
15545059 500 g 1 unit
15595049 10 g 1 unit
15575049 2 g 1 unit
15575059 5 kg 1 unit
15585049 5 g 1 unit
15545189 10 kg 1 unit
15555059 1 kg 1 unit
15565059 2 kg 1 unit
15515059 50 g 1 unit
15505059 20 g 1 unit
13 options
Product Code. 15525059
Only null left
Add to basket
Add to basket
Calibrate masses of class II and III weighing instruments. Fisherbrand™ Class F1 Stainless Steel Cylindrical Calibration Weights are intended for use in the verification or calibration of class F2 weights and for use with weighing instruments of special accuracy.
  • Class F1 stainless steel calibration weight
  • In compliance with OIML R111
Specifications
Classification F1
Material Stainless Steel
Weight (Metric) 100 g
Mass 100 g
Shape Cylindrical