missing translation for 'onlineSavingsMsg'
Learn More
Learn More
INCENP Antibody (3D2) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
1800.00 NOK - 5145.00 NOK
Specifications
| Antigen | INCENP |
|---|---|
| Clone | 3D2 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000, CyTOF-ready |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18758043
|
Novus Biologicals
NBP2-37471 |
0.1 mL |
5145.00 NOK
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18665575
|
Novus Biologicals
NBP2-37471-0.025ml |
0.025 mL |
1800.00 NOK
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
INCENP Monoclonal specifically detects INCENP in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, CyTOF-ready.Specifications
| INCENP | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| binds and activates aurora-B and -C in vivo and in vitro, chromosomal passenger protein, FLJ31633, inner centromere protein, inner centromere protein antigens (135kD, 155kD), inner centromere protein antigens 135/155kDa, inner centromere protein INCENP, MGC111393 | |
| INCENP | |
| IgG1 | |
| Affinity Purified |
| 3D2 | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000, CyTOF-ready | |
| Monoclonal | |
| Mouse | |
| Cell Cycle and Replication, Chromatin Research, DNA Repair, Mitotic Regulators, Protein Kinase | |
| PBS with 0.05% Sodium Azide | |
| 3619 | |
| Purified recombinant fragment of human INCENP (AA: 369-583) expressed in E. Coli. | |
| Primary | |
| 105.4 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title