missing translation for 'onlineSavingsMsg'
Learn More
Learn More
INCENP Antibody (3D2) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NBP2-37471-0.025ml
This item is not returnable.
View return policy
Description
INCENP Monoclonal specifically detects INCENP in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, CyTOF-ready.
Specifications
| INCENP | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000, CyTOF-ready | |
| binds and activates aurora-B and -C in vivo and in vitro, chromosomal passenger protein, FLJ31633, inner centromere protein, inner centromere protein antigens (135kD, 155kD), inner centromere protein antigens 135/155kDa, inner centromere protein INCENP, MGC111393 | |
| Mouse | |
| 105.4 kDa | |
| 0.025 mL | |
| Cell Cycle and Replication, Chromatin Research, DNA Repair, Mitotic Regulators, Protein Kinase | |
| 3619 | |
| Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles. |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 3D2 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| INCENP | |
| Purified recombinant fragment of human INCENP (AA: 369-583) expressed in E. Coli. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction