missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ProcartaPlex™ Human Immune Monitoring Panel for MAGPIX, 65plex

Product Code. 15914274
missing translation for 'orderingAttributeHoverText'
Quantity:
96 Tests
missing translation for 'unitSize'
Pack of 96
This item is not returnable. View return policy

Product Code. 15914274

missing translation for 'mfr': Invitrogen™ EPX65016500901

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The ProcartaPlex Human Immune Monitoring Panel for MAGPIX 65plex is a preconfigured multiplex immunoassay kit that measures 65 cytokines, chemokines, and growth factors using Luminex xMAP technology.

The ProcartaPlex Human Immune Monitoring Panel for MAGPIX 65plex is a preconfigured multiplex immunoassay kit that measures 65 cytokines, chemokines, and growth factors using Luminex xMAP technology. Analyze these 65 targets simultaneously for efficient immune response profiling, biomarker discovery, and validation. This kit consists of two separate kits: a 43-plex and a 22-plex kit that have been specifically designed to be used on the MAGPIX instrument that is capable of detecting up to 50 targets simultaneously. Luminex 100 and FLEXMAP 3D users should use the Human Immune Monitoring Panel 65plex (Cat. No. EPX650-10065-901) that contains all 65 targets in a single panel. The panels (43-plex and 22-plex) are provided in a ready-to-use format with individual vials of capture and detection reagents formulated at 1X concentration, requiring less pipetting and experimental setup.

ProcartaPlex preconfigured panels are extensively tested for analyte combinability, interference, and cross-reactivity to provide the highest level of validation and precision. All ProcartaPlex panels are supplied with the necessary reagents to perform the assay.

ProcartaPlex multiplex panels are available in multiple formats across six species (human, mouse, rat, nonhuman primate, porcine, and canine).

Target list [bead region]:

  • 22-plex: APRIL [52], BAFF [65], BLC (CXCL13) [15], CD30 [34], ENA-78 (CXCL5) [55], Eotaxin-2 (CCL24) [37], Eotaxin-3 (CCL26) [77], FGF-2 [75], Fractalkine (CX3CL1) [39], IL-16 [47], IL-2R (CD25) [22], IL-20 [26], I-TAC (CXCL11) [46], MCP-2 (CCL8) [18], MCP-3 (CCL7) [66], MDC (CCL22) [30], MIF [67], MIG (CXCL9) [54], TNF-RII [45], TRAIL (CD253) [29], TSLP [61], TWEAK [35]
  • 43-plex: CD40L (CD154) [74], Eotaxin (CCL11) [33], Gro-alpha (CXCL1) [61], G-CSF (CSF-3) [42], GM-CSF [44], HGF [46], IFN alpha [48], IP-10 (CXCL10) [22], IFN gamma [43], IL-1 alpha [62], IL-1 beta [18], IL-2 [19], IL-3 [73], IL-4 [20], IL-5 [21], IL-6 [25], IL-7 [26], IL-8 (CXCL8) [27], IL-9 [52], IL-10 [28], IL-12p70 [34], IL-13 [35], IL-15 [65], IL-17A (CTLA-8) [36], IL-18 [66], IL-21 [72], IL-22 [76], IL-23 [63], IL-27 [14], IL-31 [37], LIF [15], M-CSF [67], MCP-1 (CCL2) [51], MIP-1 alpha (CCL3) [12], MIP-1 beta (CCL4) [47], MIP-3 alpha (CCL20) [56], MMP-1 [64], NGF beta [55], SDF-1 alpha (CXCL12) [13], SCF [39], TNF alpha [45], TNF beta [54], VEGF-A [78]

About ProcartaPlex assays for the Luminex platform

ProcartaPlex immunoassays are based on the principles of a sandwich ELISA, using two highly specific antibodies binding to different epitopes of one protein to quantitate all protein targets simultaneously using a Luminex instrument. ProcartaPlex multiplex assays require as little as 25 μL of plasma or serum, or 50 μL of cell culture supernatant, and just four hours to obtain analyzed results.

  • Reproducible, reliable results—validated as a panel to the highest industry standard, including protein target combinability and cross-reactivity testing
  • More results per sample—measure multiple protein targets simultaneously in a single 25–50 μL sample
  • Well-established Luminex technology—highly referenced multiplexing platform for protein detection and quantitation

ProcartaPlex assays utilize Luminex xMAP (multianalyte profiling) technology for the simultaneous detection and quantitation of up to 80 protein targets in a single 25 to 50 μL sample—from plasma, serum, cell culture supernatants, and other bodily fluids.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Multiplex Panel
Quantity 96 Tests
Species Human
Combinability Non-Combinable
Detection Method Fluorescence
Assay Sensitivity See Certificate of Analysis
Assay Range See Certificate of Analysis
Format Multiplex Kit
For Use With (Equipment) Luminex™ Instruments
Product Line ProcartaPlex
Sample Type Serum, Plasma, Cell Culture Supernatants
Sample Volume Serum, Plasma: 25 μL, CCS: 50 μL
Content And Storage

• 4 vials Human Standard Mix M (lyophilized)
• 1 vial Capture Bead Mix M1 (1X)
• 1 vial Capture Bead Mix M2 (1X)
• 1 vial Biotinylated Detection Antibody Mix M1 (1X)
• 1 vial Biotinylated Detection Antibody Mix M2 (1X)
• 4 vials Human Control-High
• 4 vials Human Control-Low
• 1 Bottle Reading Buffer (1X)
• 1 Bottle Wash Buffer (10X)
• 1 Bottle Streptavidin-PE (1X)
• 1 Bottle Universal Assay Buffer (1X)
• 8-tube strip
• Adhesive film
• 2 Flat-bottom 96-well plate, black

Store at 2°C to 8°C

Shipping Condition Wet Ice
Research Area Immunology
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.