Learn More
Ebro Electronic™ Thermocouple Rod Probes
GREENER_CHOICE
Product Code. 10464473
Change view
Click to view available options
Length (Metric) Probe:
185 mm
300 mm
Diameter (Metric) Probe:
0.5 mm
1.5 mm
3 mm
Model:
TPN 100
TPN 120
TPN 120-30
TPN 140
TPN 140-30
Unit Size:
Each
Product Code. Length (Metric) Probe Diameter (Metric) Probe Model unitSize
10464473 185 mm 3 mm TPN 140 Each
11703489 185 mm 0.5 mm TPN 100 Each
10583821 185 mm 1.5 mm TPN 120 Each
10289969 300 mm 1.5 mm TPN 120-30 Each
10167999 300 mm 3 mm TPN 140-30 Each
5 options
Product Code. 10464473
Brand: Ebro Electronic™ 13410607
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Rod probes for temperature measurement of air, ovens, fluid and gases.
For various applications in food, laboratory, industial use.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance RoHS Directive 2011/65/EU
Diameter (Metric) Probe 3 mm
Disposable Single-use
For Use With (Application) Measurement of air, oven, fluids and gas temperatures
Height (Metric) 15 mm
Immersion Type Total or Partial
Length (Metric) 220 mm
Length (Metric) Probe 185 mm
Material Metal
Measurement Time 4 sec.
Model TPN 140
Probe Type NiCrNi rod
Weight (Metric) 0.24 kg
Product Type Immersion Probe
Max. Temperature (Metric) 1100°C
Min. Temperature (Metric) -40°C
Temperature Range (English) -40 °F to +2012 °F
Temperature Range (Metric) -40°C to +1100°C
For Use With (Equipment) TFN 520
Accuracy DIN IEC 584, Class 2
Width (Metric) 15 mm