Learn More
Ebro Electronic™ Thermocouple Rod Probes
GREENER_CHOICE
Product Code. 10167999
Click to view available options
Length (Metric) Probe:
185 mm
300 mm
Diameter (Metric) Probe:
0.5 mm
1.5 mm
3 mm
Model:
TPN 100
TPN 120
TPN 120-30
TPN 140
TPN 140-30
Unit Size:
Each
Product Code. 10167999
Brand: Ebro Electronic™ 13410415
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Rod probes for temperature measurement of air, ovens, fluid and gases.
For various applications in food, laboratory, industial use.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance RoHS Directive 2011/65/EU
Diameter (Metric) Probe 3 mm
Disposable Single-use
For Use With (Application) Basic rod probes, Temperature measurement of air, ovens, fluids and gases.
Height (Metric) 15 mm
Length (Metric) 300 mm
Length (Metric) Probe 300 mm
Model TPN 140-30
Probe Type NiCrNi rod
Weight (Metric) 15 mm
Product Type Rod Probe
Temperature Range (Metric) -40°C to 1100°C
For Use With (Equipment) TFN 520
Width (Metric) 0.12 kg