Learn More
Cytiva Sephadex™ G-50 and G-100 DNA Grade Agarose Gels
Product Code. 11574925 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
25 g
100 g
Bed Volume:
15 to 20 mL/g in Distilled Water
9 to 11 mL/g in Distilled Water
Particle Size:
20 to 50 μm
50 to 150 μm
Unit Size:
100g
25g
Product Code. Quantity Bed Volume Particle Size unitSize
11574925 25 g 9 to 11 mL/g in Distilled Water 50 to 150 μm 25g
11584925 100 g 9 to 11 mL/g in Distilled Water 50 to 150 μm 100g
11594925 100 g 15 to 20 mL/g in Distilled Water 20 to 50 μm 100g
3 options
Product Code. 11574925
Brand: Cytiva 17057301
Used for purification of DNA from small molecules. Cytiva illustra™ Sephadex™ G-50 and G-100 DNA Grade Agarose Gels purify DNA by gel filtration.
  • Tested to ensure reproducibly high recovery of DNA
  • Suitable for researchers who prefer to prepare their own columns for purifying nucleic acids
TRUSTED_SUSTAINABILITY
Specifications
Quantity 25 g
Particle Size 50 to 150 μm
For Use With (Equipment) Auto Seq G-50 column, ProbeQuant G-50 micro column, MicroSpin G-50 column and NICK column
Bed Volume 9 to 11 mL/g in Distilled Water
Product Type G-50 and G-100 DNA Grade Agarose Gels
Format Beaded Gel
For Use With (Application) For purification of DNA from small molecules by gel filtration
pH Range 2 to 10
Support Type Cross-linked Dextran with Epichlorohydrin
Sample Type DNA
Content And Storage 4° to 30°C