missing translation for 'onlineSavingsMsg'
Learn More

IL37 Mouse anti-Human, Clone: 7F1A1, Proteintech

Mouse Monoclonal Antibody

Brand:  Proteintech 60296-1-IG-20UL

 View more versions of this product

Product Code. 16847375

  • 1710.33 NOK / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The protein encoded by this gene is a member of the interleukin 1 cytokine family. This cytokine can bind to, and may be a ligand for interleukin 18 receptor (IL18R1/IL-1Rrp). This cytokine also binds to interleukin 18 binding protein (IL18BP), an inhibitory binding protein of interleukin 18 (IL18), and subsequently forms a complex with IL18 receptor beta subunit, and through which it inhibits the activity of IL18. This gene along with eight other interleukin 1 family genes form a cytokine gene cluster on chromosome 2. Five alternatively spliced transcript variants encoding distinct isoforms have been reported.
Specifications

Specifications

IL37
Monoclonal
1.5 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9NZH6
IL37
IL37 Fusion Protein Ag2441
20 μL
Primary
Human
Antibody
IgG2a
Western Blot, Immunohistochemistry (Paraffin)
7F1A1
Unconjugated
IL37
FIL1, FIL1 zeta, FIL1(ZETA), FIL1Z, IL 1 zeta, IL 1F7, IL 1H4, IL 1RP1, IL 1X, IL1F7, IL1H4, IL1RP1, IL37, Interleukin 1 family member 7, Interleukin 1 homolog 4, Interleukin 1 related protein, Interleukin 1 zeta
Mouse
Protein G
RUO
27178
-20°C
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.