missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
The protein encoded by this gene is a member of the interleukin 1 cytokine family. This cytokine can bind to, and may be a ligand for interleukin 18 receptor (IL18R1/IL-1Rrp). This cytokine also binds to interleukin 18 binding protein (IL18BP), an inhibitory binding protein of interleukin 18 (IL18), and subsequently forms a complex with IL18 receptor beta subunit, and through which it inhibits the activity of IL18. This gene along with eight other interleukin 1 family genes form a cytokine gene cluster on chromosome 2. Five alternatively spliced transcript variants encoding distinct isoforms have been reported.
Specifica
Specifica
| Antigen | IL37 |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | 7F1A1 |
| Concentration | 1.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | IL37 |
| Gene Accession No. | Q9NZH6 |
| Gene Alias | FIL1, FIL1 zeta, FIL1(ZETA), FIL1Z, IL 1 zeta, IL 1F7, IL 1H4, IL 1RP1, IL 1X, IL1F7, IL1H4, IL1RP1, IL37, Interleukin 1 family member 7, Interleukin 1 homolog 4, Interleukin 1 related protein, Interleukin 1 zeta |
| Vedi altri risultati |
Titolo del prodotto
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.
Individuate un'opportunità di miglioramento?