Learn More
IL-12B Mouse anti-Human, Unconjugated, Clone: 4B2A8, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 66288-1-IG-150UL
 View more versions of this product
Product Code. 16848025
  • 4836.10 NOK / 150µL
Description
The IL-12B antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human IL-12B. This antibody recognizes human, pig antigen. The IL-12B antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
IL-12B
Monoclonal
1373 μg/mL
Mouse
Primary
Human, Pig
Western Blot, ELISA
4B2A8
Unconjugated
150 μL
3593
IgG2a