missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-12B Mouse anti-Human, Unconjugated, Clone: 4B2A8, Proteintech
Mouse Monoclonal Antibody
1710.33 NOK - 4836.10 NOK
Specifications
| Antigen | IL-12B |
|---|---|
| Clone | 4B2A8 |
| Concentration | 1373 μg/mL |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16858025
|
Proteintech
66288-1-IG-20UL |
20 μL |
1710.33 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16848025
|
Proteintech
66288-1-IG-150UL |
150 μL |
4836.10 NOK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The IL-12B antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human IL-12B. This antibody recognizes human, pig antigen. The IL-12B antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| IL-12B | |
| 1373 μg/mL | |
| Monoclonal | |
| Mouse | |
| 3593 | |
| Primary |
| 4B2A8 | |
| Western Blot, ELISA | |
| Unconjugated | |
| Human, Pig | |
| IgG2a |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title