Learn More
IL-12B Mouse anti-Human, Unconjugated, Clone: 4B2A8, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66288-1-IG-20UL
This item is not returnable.
View return policy
Description
The IL-12B antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human IL-12B. This antibody recognizes human, pig antigen. The IL-12B antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| IL-12B | |
| Monoclonal | |
| 1373 μg/mL | |
| Mouse | |
| Primary | |
| Human, Pig |
| Western Blot, ELISA | |
| 4B2A8 | |
| Unconjugated | |
| 20 μL | |
| 3593 | |
| IgG2a |