missing translation for 'onlineSavingsMsg'
Learn More

Proteintech IHCeasy CD43 Ready-To-Use IHC Kit

Product Code. 17436723 Shop All Proteintech Products
Change view
Click to view available options
Packaging Quantity:
50 Tests
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Packaging Quantity unitSize
17436723 50 Tests 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17436723 Supplier Proteintech Supplier No. KHC001550T

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A ready-to-use IHC kit for the staining of CD43.

  • This kit provides all reagents, from antigen retrieval to cover slip mounting, that require little to no diluting or handling prior to use.
  • Simply apply the reagents to your sample slide according to the protocol and you're steps away from obtaining high-quality IHC data.

Specifications

Target Species Human
Sample Type FFPE Tissue
Content And Storage Store at 2°C to 8°C
Packaging Quantity 50 Tests
Sufficient For 50 Reactions
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.