Learn More
Hydrazine monohydrobromide, 98%, Thermo Scientific™
Product Code. 11494037
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
Product Code. 11494037
Brand: Thermo Scientific Alfa Aesar A19462.18
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 13775-80-9
Molecular Formula BrH5N2
Molecular Weight (g/mol) 112.958
MDL Number MFCD00065394
InChI Key DVHXJLRODLTJOD-UHFFFAOYSA-N
Synonym hydrazine monohydrobromide, hydrazine hydrobromide, hydrazine, monohydrobromide, hydrazinium bromide, hydrobromic acid hydrazine, hydrazine, hydrobromide 1:1, hydrazine mono hbr, diamine hydrobromide, acmc-1bt6e, hydrazine hydrobromide, mono
PubChem CID 159693
IUPAC Name hydrazine;hydrobromide
SMILES NN.Br
Specifications
Boiling Point ≈190°C (dec.)
Melting Point 92° to 93°C
Quantity 50 g
UN Number UN2928
Formula Weight 112.96
Percent Purity 98%
Chemical Name or Material Hydrazine monohydrobromide