accessibility menu, dialog, popup

UserName

CAS RN 13775-80-9

HBrNH2H2N

CAS RN 13775-80-9

IUPAC Name: hydrogen hydrazine bromide
Synonyms: hydrogen hydrazine bromide
Molecular Weight (g/mol): 112.96
Molecular Formula: BrH5N2
InChi Key: DVHXJLRODLTJOD-UHFFFAOYSA-N
SMILES: [H+].[Br-].NN

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results