Learn More
Abnova™ Human ZNF322A Full-length ORF (NP_078915.2, 1 a.a. - 402 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00079692-P01.10ug
Additional Details : Weight : 0.00010kg
Description
ZNF322A is a member of the zinc-finger transcription factor family and may regulate transcriptional activation in MAPK (see MAPK1; MIM 176948) signaling pathways (Li et al., 2004 [PubMed 15555580]).[supplied by OMIM]
Sequence: MYTSEEKCNQRTQKRKIYNVCPRKGKKIFIHMHEIIQIDGHIYQCLECKQNFCENLALIMCERTHTGEKPYKCDMCEKTFVQSSDLTSHQRIHNYEKPYKCSKCEKSFWHHLALSGHQRTHAGKKFYTCDICGKNFGQSSDLLVHQRSHTGEKPYLCSECDKCFSRSTNLIRHRRTHTGEKPFKCLECEKAFSGKSDLISHQRTHTGERPYKCNKCEKSYRHRSAFIVHKRVHTGEKPYKCGACEKCFGQKSDLIVHQRVHTGEKPYKCLECMRSFTRSANLIRHQATHTHTFKCLEYEKSFNCSSDLIVHQRIHMEEKPHQWSACESGFLLGMDFVAQQKMRTQTEELHYKYTVCDKSFHQSSALLQHQTVHIGEKPFVCNVSEKGLELSPPHASEASQMSSpecifications
NP_078915.2 | |
Liquid | |
79692 | |
ZNF322A (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue | |
MYTSEEKCNQRTQKRKIYNVCPRKGKKIFIHMHEIIQIDGHIYQCLECKQNFCENLALIMCERTHTGEKPYKCDMCEKTFVQSSDLTSHQRIHNYEKPYKCSKCEKSFWHHLALSGHQRTHAGKKFYTCDICGKNFGQSSDLLVHQRSHTGEKPYLCSECDKCFSRSTNLIRHRRTHTGEKPFKCLECEKAFSGKSDLISHQRTHTGERPYKCNKCEKSYRHRSAFIVHKRVHTGEKPYKCGACEKCFGQKSDLIVHQRVHTGEKPYKCLECMRSFTRSANLIRHQATHTHTFKCLEYEKSFNCSSDLIVHQRIHMEEKPHQWSACESGFLLGMDFVAQQKMRTQTEELHYKYTVCDKSFHQSSALLQHQTVHIGEKPFVCNVSEKGLELSPPHASEASQMS | |
RUO | |
ZNF322A | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
73.3kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ23393/HCG12/ZNF322/ZNF388/ZNF489/bA457M11.2/bA457M11.3 | |
ZNF322A | |
Yes | |
wheat germ expression system |