missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human ZNF322A Full-length ORF (NP_078915.2, 1 a.a. - 402 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4115.00 NOK - 6245.00 NOK
Specifications
Accession Number | NP_078915.2 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 79692 |
Molecular Weight (g/mol) | 73.3kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16144353
|
Abnova™
H00079692-P01.25UG |
25 ug |
6245.00 NOK
25µg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16134353
|
Abnova™
H00079692-P01.10UG |
10 ug |
4115.00 NOK
10µg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
ZNF322A is a member of the zinc-finger transcription factor family and may regulate transcriptional activation in MAPK (see MAPK1; MIM 176948) signaling pathways (Li et al., 2004 [PubMed 15555580]).[supplied by OMIM]
Sequence: MYTSEEKCNQRTQKRKIYNVCPRKGKKIFIHMHEIIQIDGHIYQCLECKQNFCENLALIMCERTHTGEKPYKCDMCEKTFVQSSDLTSHQRIHNYEKPYKCSKCEKSFWHHLALSGHQRTHAGKKFYTCDICGKNFGQSSDLLVHQRSHTGEKPYLCSECDKCFSRSTNLIRHRRTHTGEKPFKCLECEKAFSGKSDLISHQRTHTGERPYKCNKCEKSYRHRSAFIVHKRVHTGEKPYKCGACEKCFGQKSDLIVHQRVHTGEKPYKCLECMRSFTRSANLIRHQATHTHTFKCLEYEKSFNCSSDLIVHQRIHMEEKPHQWSACESGFLLGMDFVAQQKMRTQTEELHYKYTVCDKSFHQSSALLQHQTVHIGEKPFVCNVSEKGLELSPPHASEASQMSSpecifications
NP_078915.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
73.3kDa | |
Glutathione Sepharose 4 Fast Flow | |
MYTSEEKCNQRTQKRKIYNVCPRKGKKIFIHMHEIIQIDGHIYQCLECKQNFCENLALIMCERTHTGEKPYKCDMCEKTFVQSSDLTSHQRIHNYEKPYKCSKCEKSFWHHLALSGHQRTHAGKKFYTCDICGKNFGQSSDLLVHQRSHTGEKPYLCSECDKCFSRSTNLIRHRRTHTGEKPFKCLECEKAFSGKSDLISHQRTHTGERPYKCNKCEKSYRHRSAFIVHKRVHTGEKPYKCGACEKCFGQKSDLIVHQRVHTGEKPYKCLECMRSFTRSANLIRHQATHTHTFKCLEYEKSFNCSSDLIVHQRIHMEEKPHQWSACESGFLLGMDFVAQQKMRTQTEELHYKYTVCDKSFHQSSALLQHQTVHIGEKPFVCNVSEKGLELSPPHASEASQMS | |
RUO | |
ZNF322A | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
79692 | |
ZNF322A (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ23393/HCG12/ZNF322/ZNF388/ZNF489/bA457M11.2/bA457M11.3 | |
ZNF322A | |
Recombinant | |
wheat germ expression system |