missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ZNF239 Full-length ORF (AAH26030.1, 1 a.a. - 458 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00008187-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16142302

  • 4115.00 NOK / 10µg
Estimated Shipment: 25-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

MOK2 proteins are DNA- and RNA-binding proteins that are mainly associated with nuclear RNP components, including the nucleoli and extranucleolar structures (Arranz et al., 1997 [PubMed 9121460]).[supplied by OMIM]

Sequence: MASTITGSQDCIVNHRGEVDGEPELDISPCQQWGEASSPISRNRDSVMTLQSGCFENIESETYLPLKVSSQIDTQDSSVKFCKNEPQDHQESRRLFVMEESTERKVIKGESCSENLQVKLVSDGQELASPLLNGEATCQNGQLKESLDPIDCNCKDIHGWKSQVVSCSQQRAHTEEKPCDHNNCGKILNTSPDGHPYEKIHTAEKQYECSQCGKNFSQSSELLLHQRDHTEEKPYKCEQCGKGFTRSSSLLIHQAVHTDEKPYKCDKCGKGFTRSSSLLIHHAVHTGEKPYKCDKCGKGFSQSSKLHIHQRVHTGEKPYECEECGMSFSQRSNLHIHQRVHTGERPYKCGECGKGFSQSSNLHIHRCIHTGEKPYQCYECGKGFSQSSDLRIHLRVHTGEKPYHCGKCGKGFSQSSKLLIHQRVHTGEKPYECSKCGKGFSQSSNLHIHQRVHKKDPR
Specifications

Specifications

AAH26030.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
78kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
HOK-2/MOK2
ZNF239
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
8187
ZNF239 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MASTITGSQDCIVNHRGEVDGEPELDISPCQQWGEASSPISRNRDSVMTLQSGCFENIESETYLPLKVSSQIDTQDSSVKFCKNEPQDHQESRRLFVMEESTERKVIKGESCSENLQVKLVSDGQELASPLLNGEATCQNGQLKESLDPIDCNCKDIHGWKSQVVSCSQQRAHTEEKPCDHNNCGKILNTSPDGHPYEKIHTAEKQYECSQCGKNFSQSSELLLHQRDHTEEKPYKCEQCGKGFTRSSSLLIHQAVHTDEKPYKCDKCGKGFTRSSSLLIHHAVHTGEKPYKCDKCGKGFSQSSKLHIHQRVHTGEKPYECEECGMSFSQRSNLHIHQRVHTGERPYKCGECGKGFSQSSNLHIHRCIHTGEKPYQCYECGKGFSQSSDLRIHLRVHTGEKPYHCGKCGKGFSQSSKLLIHQRVHTGEKPYECSKCGKGFSQSSNLHIHQRVHKKDPR
RUO
ZNF239
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers