missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human ZNF239 Full-length ORF (AAH26030.1, 1 a.a. - 458 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4115.00 NOK - 6245.00 NOK
Specifications
Accession Number | AAH26030.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 8187 |
Molecular Weight (g/mol) | 78kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16152302
|
Abnova™
H00008187-P01.25UG |
25 ug |
6245.00 NOK
25µg |
Estimated Shipment: 24-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16142302
|
Abnova™
H00008187-P01.10UG |
10 ug |
4115.00 NOK
10µg |
Estimated Shipment: 24-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
MOK2 proteins are DNA- and RNA-binding proteins that are mainly associated with nuclear RNP components, including the nucleoli and extranucleolar structures (Arranz et al., 1997 [PubMed 9121460]).[supplied by OMIM]
Sequence: MASTITGSQDCIVNHRGEVDGEPELDISPCQQWGEASSPISRNRDSVMTLQSGCFENIESETYLPLKVSSQIDTQDSSVKFCKNEPQDHQESRRLFVMEESTERKVIKGESCSENLQVKLVSDGQELASPLLNGEATCQNGQLKESLDPIDCNCKDIHGWKSQVVSCSQQRAHTEEKPCDHNNCGKILNTSPDGHPYEKIHTAEKQYECSQCGKNFSQSSELLLHQRDHTEEKPYKCEQCGKGFTRSSSLLIHQAVHTDEKPYKCDKCGKGFTRSSSLLIHHAVHTGEKPYKCDKCGKGFSQSSKLHIHQRVHTGEKPYECEECGMSFSQRSNLHIHQRVHTGERPYKCGECGKGFSQSSNLHIHRCIHTGEKPYQCYECGKGFSQSSDLRIHLRVHTGEKPYHCGKCGKGFSQSSKLLIHQRVHTGEKPYECSKCGKGFSQSSNLHIHQRVHKKDPRSpecifications
AAH26030.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
78kDa | |
Glutathione Sepharose 4 Fast Flow | |
MASTITGSQDCIVNHRGEVDGEPELDISPCQQWGEASSPISRNRDSVMTLQSGCFENIESETYLPLKVSSQIDTQDSSVKFCKNEPQDHQESRRLFVMEESTERKVIKGESCSENLQVKLVSDGQELASPLLNGEATCQNGQLKESLDPIDCNCKDIHGWKSQVVSCSQQRAHTEEKPCDHNNCGKILNTSPDGHPYEKIHTAEKQYECSQCGKNFSQSSELLLHQRDHTEEKPYKCEQCGKGFTRSSSLLIHQAVHTDEKPYKCDKCGKGFTRSSSLLIHHAVHTGEKPYKCDKCGKGFSQSSKLHIHQRVHTGEKPYECEECGMSFSQRSNLHIHQRVHTGERPYKCGECGKGFSQSSNLHIHRCIHTGEKPYQCYECGKGFSQSSDLRIHLRVHTGEKPYHCGKCGKGFSQSSKLLIHQRVHTGEKPYECSKCGKGFSQSSNLHIHQRVHKKDPR | |
RUO | |
ZNF239 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
8187 | |
ZNF239 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HOK-2/MOK2 | |
ZNF239 | |
Recombinant | |
wheat germ expression system |