missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ZHX3 Partial ORF (NP_055850.1, 863 a.a. - 955 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00023051-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16134386

  • 4115.00 NOK / 10µg
Estimated Shipment: 11-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes a member of the zinc fingers and homeoboxes (ZHX) gene family. The encoded protein contains two C2H2-type zinc fingers and five homeodomains and forms a dimer with itself or with zinc fingers and homeoboxes family member 1. In the nucleus, the dimerized protein interacts with the A subunit of the ubiquitous transcription factor nuclear factor-Y and may function as a transcriptional repressor. [provided by RefSeq]

Sequence: EDLQNLCDKTQMSSQQVKQWFAEKMGEETRAVADTGSEDQGPGTGELTAVHKGMGDTYSEVSENSESWEPRVPEASSEPFDTSSPQAGRQLET
Specifications

Specifications

NP_055850.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
35.97kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
EDLQNLCDKTQMSSQQVKQWFAEKMGEETRAVADTGSEDQGPGTGELTAVHKGMGDTYSEVSENSESWEPRVPEASSEPFDTSSPQAGRQLET
RUO
ZHX3
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, Protein Array, ELISA, Western Blot
23051
ZHX3 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
KIAA0395/TIX1
ZHX3
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers