missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human ZHX3 Partial ORF (NP_055850.1, 863 a.a. - 955 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4115.00 NOK - 6245.00 NOK
Specifications
Accession Number | NP_055850.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 23051 |
Molecular Weight (g/mol) | 35.97kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16144386
|
Abnova™
H00023051-Q01.25UG |
25 ug |
6245.00 NOK
25µg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16134386
|
Abnova™
H00023051-Q01.10UG |
10 ug |
4115.00 NOK
10µg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes a member of the zinc fingers and homeoboxes (ZHX) gene family. The encoded protein contains two C2H2-type zinc fingers and five homeodomains and forms a dimer with itself or with zinc fingers and homeoboxes family member 1. In the nucleus, the dimerized protein interacts with the A subunit of the ubiquitous transcription factor nuclear factor-Y and may function as a transcriptional repressor. [provided by RefSeq]
Sequence: EDLQNLCDKTQMSSQQVKQWFAEKMGEETRAVADTGSEDQGPGTGELTAVHKGMGDTYSEVSENSESWEPRVPEASSEPFDTSSPQAGRQLETSpecifications
NP_055850.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
35.97kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
KIAA0395/TIX1 | |
ZHX3 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
23051 | |
ZHX3 (Human) Recombinant Protein (Q01) | |
EDLQNLCDKTQMSSQQVKQWFAEKMGEETRAVADTGSEDQGPGTGELTAVHKGMGDTYSEVSENSESWEPRVPEASSEPFDTSSPQAGRQLET | |
RUO | |
ZHX3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |