Learn More
R&D Systems™ Human Wnt-8a (NP_490645.1) VersaClone cDNA
Specifications
Specifications
| Gene | Wnt-8 A |
| Accession Number | NP_490645.1 |
| Gene ID (Entrez) | 7478 |
| Gene Alias | protein Wnt-8 A, Protein Wnt-8 d, wingless-type MMTV integration site family, member 8 A, WNT8DWNT8d |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Quantity | 10 μg |