Learn More
R&D Systems™ Human Wnt-7b (NP_478679.1) VersaClone cDNA
Specifications
Specifications
| Gene | Wnt-7 b |
| Accession Number | NP_478679.1 |
| Gene ID (Entrez) | 7477 |
| Gene Alias | protein Wnt-7 b, wingless-type MMTV integration site family, member 7 B |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Quantity | 10 μg |