Learn More
R&D Systems™ Human Wnt-3 (NP_110380.1) VersaClone cDNA
Specifications
Specifications
| Gene | Wnt3 |
| Accession Number | NP_110380.1 |
| Gene ID (Entrez) | 7473 |
| Gene Alias | INT4Proto-oncogene Int-4 homolog, MGC131950, MGC138321, MGC138323, proto-oncogene Wnt-3, wingless-type MMTV integration site family, member 3, WNT-3 proto-oncogene protein |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Product Line | R&D Systems |
| Quantity | 10 μg |