Learn More
Human TTLL12 knockout HEK-293T cell lysate, 1 Kit
Product Code. 32143738 Shop All Abcam Products
Product Code. 32143738 Supplier Abcam Supplier No. ab2634011Kit
 Restricted Item
Estimated Shipment: 11-11-2026
Human TTLL12 knockout HEK-293T cell/tissue lysate.
  • Species: Human
  • Tissue: TTLL12 knockout HEK-293T