Learn More
R&D Systems™ Human TSLP DuoSet ELISA
missing translation for 'orderingAttributeHoverText'
Format:
15 96-well Plates
5 96-well Plates
missing translation for 'unitSize'
1 set
Pack of 5
Description
The Human TSLP DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human TSLP.
Specifications
Specifications
| Product Type | ELISA Kit |
| Quantity | 1 Kit |
| Species | Human |
| Conjugate | Biotin |
| Assay Range | 31.20 to 2000 pg/mL |
| Cross Reactivity | <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules |
| Format | 15 96-well Plates |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 85480 |
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |