Learn More
R&D Systems™ Human Total MMP-8 Quantikine ELISA Kit
Product Code. 30000766
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
Pack of 96
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30000766 1 Kit Pack of 96
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30000766 Supplier R&D Systems™ Supplier No. DMP800B
Human Total MMP-8 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
The Human Total MMP-8 Quantikine ELISA Kit quantifies human MMP-8 in cell culture supernates (50 μL), tissue lysates (50 μL), serum (10 μL), edta plasma (25 μL), platelet-poor edta plasma (50 μL), heparin plasma (25 μL), platelet-poor heparin plasma (50 μL), saliva (10 μL).
TRUSTED_SUSTAINABILITY
Specifications
Assay Range 0.16 to 10 ng/mL
Assay Sensitivity 0.058 ng/mL
Sample Type Cell Culture Supernatant, Plasma (EDTA, Heparin), Platelet-poor Plasma (EDTA, Heparin), Saliva, Serum, Tissue Lysates
Specificity Natural and Recombinant Human MMP-8
For Use With (Application) ELISA
Gene ID (Entrez) 4317
Host Species Human
Quantity 1 Kit
Sample Volume Cell Culture Supernates (50 μL),Tissue Lysates (50 μL),Serum (10 μL),EDTA Plasma (25 μL),Platelet-poor EDTA Plasma (50 μL),Heparin Plasma (25 μL),Platelet-poor Heparin Plasma (50 μL),Saliva (10 μL)
Storage Requirements Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Synonym CLG1HNC, Collagenase 2, EC 3.4.24, EC 3.4.24.34, matrix metallopeptidase 8 (neutrophil collagenase), matrix metalloproteinase 8 (neutrophil collagenase), Matrix metalloproteinase-8, MMP8, MMP-8, neutrophil collagenase, PMNL collagenase, PMNL-CL
Target MMP-8
Total Assay Time 4.5 hr.