missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human TNFAIP2 Partial ORF (NP_006282, 552 a.a. - 650 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00007127-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16196785

  • 6245.00 NOK / 25µg
Estimated Shipment: 27-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene was identified as a gene whose expression can be induced by the tumor necrosis factor alpha (TNF) in umbilical vein endothelial cells. The expression of this gene was shown to be induced by retinoic acid in a cell line expressing a oncogenic version of the retinoic acid receptor alpha fusion protein, which suggested that this gene may be a retinoic acid target gene in acute promyelocytic leukemia. [provided by RefSeq]

Sequence: YILANADTIQHFCTQHGSPATWLQPALPTLAEIIRLQDPSAIKIEVATYATCYPDFSKGHLSAILAIKGNLSNSEVKRIRSILDVSMGAQEPSRPLFSL
Specifications

Specifications

NP_006282
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.63kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
YILANADTIQHFCTQHGSPATWLQPALPTLAEIIRLQDPSAIKIEVATYATCYPDFSKGHLSAILAIKGNLSNSEVKRIRSILDVSMGAQEPSRPLFSL
RUO
TNFAIP2
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
7127
TNFAIP2 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
B94
TNFAIP2
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers