missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human TNFAIP2 Partial ORF (NP_006282, 552 a.a. - 650 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4115.00 NOK - 6245.00 NOK
Specifications
Accession Number | NP_006282 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 7127 |
Molecular Weight (g/mol) | 36.63kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16186785
|
Abnova™
H00007127-Q01.10UG |
10 ug |
4115.00 NOK
10µg |
Estimated Shipment: 13-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16196785
|
Abnova™
H00007127-Q01.25UG |
25 ug |
6245.00 NOK
25µg |
Estimated Shipment: 13-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene was identified as a gene whose expression can be induced by the tumor necrosis factor alpha (TNF) in umbilical vein endothelial cells. The expression of this gene was shown to be induced by retinoic acid in a cell line expressing a oncogenic version of the retinoic acid receptor alpha fusion protein, which suggested that this gene may be a retinoic acid target gene in acute promyelocytic leukemia. [provided by RefSeq]
Sequence: YILANADTIQHFCTQHGSPATWLQPALPTLAEIIRLQDPSAIKIEVATYATCYPDFSKGHLSAILAIKGNLSNSEVKRIRSILDVSMGAQEPSRPLFSLSpecifications
NP_006282 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
B94 | |
TNFAIP2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
7127 | |
TNFAIP2 (Human) Recombinant Protein (Q01) | |
YILANADTIQHFCTQHGSPATWLQPALPTLAEIIRLQDPSAIKIEVATYATCYPDFSKGHLSAILAIKGNLSNSEVKRIRSILDVSMGAQEPSRPLFSL | |
RUO | |
TNFAIP2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |