missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human TNF-alpha Quantikine ELISA Kit

Product Code. 30000768
Click to view available options
Quantity:
1 Pack (6 Plates)
Unit Size:
576 tests
This item is not returnable. View return policy

Product Code. 30000768

Brand: R&D Systems™ STA00D

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Human TNF-alpha Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human TNF-alpha Quantikine ELISA Kit quantifies human TNF-alpha in cell culture supernates (50 ul), serum (50 ul), plasma (50 ul).

1 Pack (6 Plates)

TRUSTED_SUSTAINABILITY

Specifications

Assay Range 15.60 - 1000 pg/mL
Assay Sensitivity 6.23 pg/mL
Sample Type Cell Culture Supernatant, Plasma (EDTA, Heparin), Serum
Specificity HIV-1 Gag p24
For Use With (Application) ELISA
Gene ID (Entrez) 7124
Host Species Human
Quantity 1 Pack (6 Plates)
Sample Volume Cell Culture Supernates (50 uL),Serum (50 uL),EDTA Plasma (50 uL),Heparin Plasma (50 uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym APC1 protein, Cachectin, Cachetin, DIF, TNF, TNF, monocyte-derived, TNF-a, TNFA, TNFalpha, TNF-alphacachectin, TNFATNF, macrophage-derived, TNFG1F, TNFSF1A, TNFSF2, TNFSF2TNF superfamily, member 2, tumor necrosis factor, tumor necrosis factor (TNF superfamily, member 2), tumor necrosis factor alpha, Tumor necrosis factor ligand superfamily member 2, tumor necrosis factor-alpha
Target TNF-alpha
Total Assay Time 4.0 hours
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.