Learn More
ACROBiosystems Human TIM-1 / HAVCR1 Protein, His Tag
Brand: ACROBiosystems KI1-H52H3-100ug
Additional Details : Weight : 0.00010kg
Description
Hepatitis A virus cellular receptor 1 is also known as HAVCR1, HAVCR, KIM1, TIM, TIM1, TIMD1, is widely expressed with highest levels in kidney and testis. The protein encoded by HAVCR1 gene is a membrane receptor for both human hepatitis A virus (HHAV) and TIMD4. The encoded protein may be involved in the moderation of asthma and allergic diseases. The reference genome represents an allele that retains a MTTVP amino acid segment that confers protection against atopy in HHAV seropositive individuals. Three transcript variants encoding the same protein have been found for this gene. HAVCR1 may play a role in T-helper cell development, the regulation of asthma and allergic diseases and in kidney injury and repair. In case of human hepatitis A virus (HHAV) infection, functions as a cell-surface receptor for the virus.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 31.4 kDa.Specifications
Powder | |
100 μg | |
Ser 21 - Gly 295 | |
HAVCR1,KIM1,TIM1,TIMD1 | |
Unconjugated | |
HEK293 | |
Human |
31.4 kDa | |
Human TIM-1 / HAVCR1, His Tag (KI1-H52H3) is expressed from human 293 cells (HEK293). It contains AA Ser 21 - Gly 295 (Accession # Q96D42-1). | |
-20°C | |
TIM-1 | |
His Tag | |
ADC |