Learn More
ACROBiosystems Human TIM-1 / HAVCR1 Protein, His Tag
5297.56 NOK - 38899.20 NOK
Specifications
Molecular Weight (g/mol) | 31.4 kDa |
---|---|
Source | Human TIM-1 / HAVCR1, His Tag (KI1-H52H3) is expressed from human 293 cells (HEK293). It contains AA Ser 21 - Gly 295 (Accession # Q96D42-1). |
Immunogen | Ser 21 - Gly 295 |
Storage Requirements | -20°C |
Gene Alias | HAVCR1,KIM1,TIM1,TIMD1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17226290
|
ACROBiosystems
KI1-H52H3-1MG |
1 mg |
38899.20 NOK
1mg |
Estimated Shipment: 11-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17216290
|
ACROBiosystems
KI1-H52H3-100UG |
100 μg |
5297.56 NOK
100µg |
Estimated Shipment: 11-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Hepatitis A virus cellular receptor 1 is also known as HAVCR1, HAVCR, KIM1, TIM, TIM1, TIMD1, is widely expressed with highest levels in kidney and testis. The protein encoded by HAVCR1 gene is a membrane receptor for both human hepatitis A virus (HHAV) and TIMD4. The encoded protein may be involved in the moderation of asthma and allergic diseases. The reference genome represents an allele that retains a MTTVP amino acid segment that confers protection against atopy in HHAV seropositive individuals. Three transcript variants encoding the same protein have been found for this gene. HAVCR1 may play a role in T-helper cell development, the regulation of asthma and allergic diseases and in kidney injury and repair. In case of human hepatitis A virus (HHAV) infection, functions as a cell-surface receptor for the virus.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 31.4 kDa.Specifications
31.4 kDa | |
Ser 21 - Gly 295 | |
HAVCR1,KIM1,TIM1,TIMD1 | |
Unconjugated | |
His Tag | |
Powder |
Human TIM-1 / HAVCR1, His Tag (KI1-H52H3) is expressed from human 293 cells (HEK293). It contains AA Ser 21 - Gly 295 (Accession # Q96D42-1). | |
-20°C | |
TIM-1 | |
Human | |
HEK293 | |
ADC |