Learn More
ACROBiosystems Human TIM-1 / HAVCR1 Protein, His Tag
Brand:  ACROBiosystems KI1-H52H3-1mg
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 17226290
  • 39223.36 NOK / 1mg
Estimated Shipment: 10-05-2024
Description
Hepatitis A virus cellular receptor 1 is also known as HAVCR1, HAVCR, KIM1, TIM, TIM1, TIMD1, is widely expressed with highest levels in kidney and testis. The protein encoded by HAVCR1 gene is a membrane receptor for both human hepatitis A virus (HHAV) and TIMD4. The encoded protein may be involved in the moderation of asthma and allergic diseases. The reference genome represents an allele that retains a MTTVP amino acid segment that confers protection against atopy in HHAV seropositive individuals. Three transcript variants encoding the same protein have been found for this gene. HAVCR1 may play a role in T-helper cell development, the regulation of asthma and allergic diseases and in kidney injury and repair. In case of human hepatitis A virus (HHAV) infection, functions as a cell-surface receptor for the virus.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 31.4 kDa.
Specifications
Specifications
31.4 kDa
Human TIM-1 / HAVCR1, His Tag (KI1-H52H3) is expressed from human 293 cells (HEK293). It contains AA Ser 21 - Gly 295 (Accession # Q96D42-1).
-20°C
TIM-1
Human
HEK293
ADC
1 mg
Ser 21 - Gly 295
HAVCR1,KIM1,TIM1,TIMD1
Unconjugated
His Tag
Powder