missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human TFPI Quantikine ELISA Kit
Click to view available options
Sufficient For:
1 x 96-well Plate
Unit Size:
1 set
Description
The Human TFPI Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human TFPI in cell culture supernates (50 ul), edta plasma (10 ul), heparin plasma (10 ul), citrate plasma (10 ul), urine (50 ul).
Specifications
Specifications
| Assay Range | 31.20 - 2000pg/mL |
| Assay Sensitivity | 6.7pg/mL |
| Conjugate | HRP |
| Sample Type | Cell Culture Supernatant, Plasma (EDTA, Citrate, Heparin), Urine |
| Specificity | Natural and Recombinant Human TFPI. This kit recognizes predominantly free TFPI and a very small percentage of the LDL- and HDL-bound TFPI. |
| Sufficient For | 1 x 96-well Plate |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 7035 |
| Sample Volume | Cell Culture Supernates (50uL),EDTA Plasma (10uL),Heparin Plasma (10uL),Citrate Plasma (10uL),Urine (50uL) |
| Storage Requirements | Store the unopened product at 2° to 8°C. Do not use past expiration date. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction