missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human Tenascin C ELISA Kit
Description
Human Tenascin C quantitates human Tenascin C in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Tenascin C.
Tenascin is a high molecular weight, multifunctional, extracellular matrix glycoprotein expressed in association with mesenchymal-epithelial interactions during development and in the neovasculature and stroma of undifferentiated tumors (Erickson and Bourdon 1989). It has been described under a variety of names: cytotactin, hexabrachion protein, J1, myotendinous antigen (MI) and glioma mesenchymal extracellular matrix (GMEM). The tenascin molecule is a disulfide-linked hexamer. The expression of tenascin is associated with development and growth, both normal and pathological, whereas the distribution in normal adult tissue is restricted.
Specifications
Specifications
| Accession Number | 150-225; AI528729; C130033P17Rik; cytotactin; deafness, autosomal dominant 56; DFNA56; Glioma-associated-extracellular matrix antigen; GMEM; GP; GP 150-225; hexabrachion; hexabrachion (tenascin); Hxb; JI; MGC144208; MGC144209; MGC167029; Myotendinous antigen; neuronectin; Ten; tenascin; tenascin C; Tenascin-C; tenascin-C additional domain 1; tenascin-C isoform 14/AD1/16; TN; Tnc; TN-C |
| Assay Range | 6.1 to 1,500 pg/mL |
| Assay Sensitivity | 6.1 pg/mL |
| Conjugate | HRP |
| Product Type | ELISA |
| Sample Type | Plasma, Serum, Supernatant |
| For Use With (Equipment) | Colorimetric Microplate Reader |
| Gene ID (Entrez) | 3371 |
| Gene Symbol | TNC |
| Interassay CV | <12% |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?