Learn More
Abnova™ Human SULT1C1 Full-length ORF (AAH05353, 1 a.a. - 296 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00006819-P01.25ug
Additional Details : Weight : 0.00010kg
Description
Sulfotransferase enzymes catalyze the sulfate conjugation of many hormones, neurotransmitters, drugs, and xenobiotic compounds. These cytosolic enzymes are different in their tissue distributions and substrate specificities. The gene structure (number and length of exons) is similar among family members. This gene encodes a protein that belongs to the SULT1 subfamily, responsible for transferring a sulfo moiety from PAPS to phenol-containing compounds. Two alternatively spliced transcript variants encoding different isoforms have been described for this gene. [provided by RefSeq]
Sequence: MALTSDLGKQIKLKEVEGTLLQPATVDNWSQIQSFEAKPDDLLICTYPKAGTTWIQEIVDMIEQNGDVEKCQRAIIQHRHPFIEWARPPQPSGVEKAKAMPSPRILKTHLSTQLLPPSFWENNCKFLYVARNAKDCMVSYYHFQRMNHMLPDPGTWEEYFETFINGKVVWGSWFDHVKGWWEMKDRHQILFLFYEDIKRDPKHEIRKVMQFMGKKVDETVLDKIVQETSFEKMKENPMTNRSTVSKSILDQSISSFMRKGTVGDWKNHFTVAQNERFDEIYRRKMEGTSINFCMELSpecifications
AAH05353 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
58.30kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ST1C1/ST1C2/SULT1C1/humSULTC2 | |
SULT1C2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
6819 | |
SULT1C1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MALTSDLGKQIKLKEVEGTLLQPATVDNWSQIQSFEAKPDDLLICTYPKAGTTWIQEIVDMIEQNGDVEKCQRAIIQHRHPFIEWARPPQPSGVEKAKAMPSPRILKTHLSTQLLPPSFWENNCKFLYVARNAKDCMVSYYHFQRMNHMLPDPGTWEEYFETFINGKVVWGSWFDHVKGWWEMKDRHQILFLFYEDIKRDPKHEIRKVMQFMGKKVDETVLDKIVQETSFEKMKENPMTNRSTVSKSILDQSISSFMRKGTVGDWKNHFTVAQNERFDEIYRRKMEGTSINFCMEL | |
RUO | |
SULT1C2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |