missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human SULT1C1 Full-length ORF (AAH05353, 1 a.a. - 296 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

4115.00 NOK - 6245.00 NOK

Specifications

Accession Number AAH05353
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 6819
Molecular Weight (g/mol) 58.30kDa
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16141142
View Documents Promotion Details
Abnova™
H00006819-P01.25UG
25 ug
6245.00 NOK
25µg
View alternative products
Save Up to 
»
Estimated Shipment: 11-06-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16131142
View Documents Promotion Details
Abnova™
H00006819-P01.10UG
10 ug
4115.00 NOK
10µg
View alternative products
Save Up to 
»
Estimated Shipment: 11-06-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Sulfotransferase enzymes catalyze the sulfate conjugation of many hormones, neurotransmitters, drugs, and xenobiotic compounds. These cytosolic enzymes are different in their tissue distributions and substrate specificities. The gene structure (number and length of exons) is similar among family members. This gene encodes a protein that belongs to the SULT1 subfamily, responsible for transferring a sulfo moiety from PAPS to phenol-containing compounds. Two alternatively spliced transcript variants encoding different isoforms have been described for this gene. [provided by RefSeq]

Sequence: MALTSDLGKQIKLKEVEGTLLQPATVDNWSQIQSFEAKPDDLLICTYPKAGTTWIQEIVDMIEQNGDVEKCQRAIIQHRHPFIEWARPPQPSGVEKAKAMPSPRILKTHLSTQLLPPSFWENNCKFLYVARNAKDCMVSYYHFQRMNHMLPDPGTWEEYFETFINGKVVWGSWFDHVKGWWEMKDRHQILFLFYEDIKRDPKHEIRKVMQFMGKKVDETVLDKIVQETSFEKMKENPMTNRSTVSKSILDQSISSFMRKGTVGDWKNHFTVAQNERFDEIYRRKMEGTSINFCMEL
Specifications

Specifications

AAH05353
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
58.30kDa
Glutathione Sepharose 4 Fast Flow
MALTSDLGKQIKLKEVEGTLLQPATVDNWSQIQSFEAKPDDLLICTYPKAGTTWIQEIVDMIEQNGDVEKCQRAIIQHRHPFIEWARPPQPSGVEKAKAMPSPRILKTHLSTQLLPPSFWENNCKFLYVARNAKDCMVSYYHFQRMNHMLPDPGTWEEYFETFINGKVVWGSWFDHVKGWWEMKDRHQILFLFYEDIKRDPKHEIRKVMQFMGKKVDETVLDKIVQETSFEKMKENPMTNRSTVSKSILDQSISSFMRKGTVGDWKNHFTVAQNERFDEIYRRKMEGTSINFCMEL
RUO
SULT1C2
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, Protein Array, ELISA, Western Blot
6819
SULT1C1 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ST1C1/ST1C2/SULT1C1/humSULTC2
SULT1C2
Recombinant
wheat germ expression system
Videos
SDS
Documents

Documents

Product Certifications