missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human STARD6 Full-length ORF (AAI46469.1, 1 a.a. - 220 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00147323-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16188453

  • 4115.00 NOK / 10µg
Estimated Shipment: 01-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Cholesterol homeostasis is regulated, at least in part, by sterol regulatory element (SRE)-binding proteins (e.g., SREBP1; MIM 184756) and by liver X receptors (e.g., LXRA; MIM 602423). Upon sterol depletion, LXRs are inactive and SREBPs are cleaved, after which they bind promoter SREs and activate genes involved in cholesterol biosynthesis and uptake. Sterol transport is mediated by vesicles or by soluble protein carriers, such as steroidogenic acute regulatory protein (STAR; MIM 600617). STAR is homologous to a family of proteins containing a 200- to 210-amino acid STAR-related lipid transfer (START) domain, including STARD6 (Soccio et al., 2002 [PubMed 12011452]).[supplied by OMIM]

Sequence: MDFKAIAQQTAQEVLGYNRDTSGWKVVKTSKKITVSSKASRKFHGNLYRVEGIIPESPAKLSDFLYQTGDRITWDKSLQVYNMVHRIDSDTFICHTITQSFAVGSISPRDFIDLVYIKRYEGNMNIISSKSVDFPEYPPSSNYIRGYNHPCGFVCSPMEENPAYSKLVMFVQTEMRGKLSPSIIEKTMPSNLVNFILNAKDGIKAHRTPSRRGFHHNSHS
Specifications

Specifications

AAI46469.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
51.15kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
STARD6
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, Protein Array, ELISA, Western Blot
147323
STARD6 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MDFKAIAQQTAQEVLGYNRDTSGWKVVKTSKKITVSSKASRKFHGNLYRVEGIIPESPAKLSDFLYQTGDRITWDKSLQVYNMVHRIDSDTFICHTITQSFAVGSISPRDFIDLVYIKRYEGNMNIISSKSVDFPEYPPSSNYIRGYNHPCGFVCSPMEENPAYSKLVMFVQTEMRGKLSPSIIEKTMPSNLVNFILNAKDGIKAHRTPSRRGFHHNSHS
RUO
STARD6
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers