Learn More
Abnova™ Human STARD6 Full-length ORF (AAI46469.1, 1 a.a. - 220 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4115.00 NOK - 6245.00 NOK
Specifications
Accession Number | AAI46469.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 147323 |
Molecular Weight (g/mol) | 51.15kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16198453
|
Abnova™
H00147323-P01.25UG |
25 ug |
6245.00 NOK
25µg |
Estimated Shipment: 19-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16188453
|
Abnova™
H00147323-P01.10UG |
10 ug |
4115.00 NOK
10µg |
Estimated Shipment: 19-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Cholesterol homeostasis is regulated, at least in part, by sterol regulatory element (SRE)-binding proteins (e.g., SREBP1; MIM 184756) and by liver X receptors (e.g., LXRA; MIM 602423). Upon sterol depletion, LXRs are inactive and SREBPs are cleaved, after which they bind promoter SREs and activate genes involved in cholesterol biosynthesis and uptake. Sterol transport is mediated by vesicles or by soluble protein carriers, such as steroidogenic acute regulatory protein (STAR; MIM 600617). STAR is homologous to a family of proteins containing a 200- to 210-amino acid STAR-related lipid transfer (START) domain, including STARD6 (Soccio et al., 2002 [PubMed 12011452]).[supplied by OMIM]
Sequence: MDFKAIAQQTAQEVLGYNRDTSGWKVVKTSKKITVSSKASRKFHGNLYRVEGIIPESPAKLSDFLYQTGDRITWDKSLQVYNMVHRIDSDTFICHTITQSFAVGSISPRDFIDLVYIKRYEGNMNIISSKSVDFPEYPPSSNYIRGYNHPCGFVCSPMEENPAYSKLVMFVQTEMRGKLSPSIIEKTMPSNLVNFILNAKDGIKAHRTPSRRGFHHNSHSSpecifications
AAI46469.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
51.15kDa | |
Glutathione Sepharose 4 Fast Flow | |
MDFKAIAQQTAQEVLGYNRDTSGWKVVKTSKKITVSSKASRKFHGNLYRVEGIIPESPAKLSDFLYQTGDRITWDKSLQVYNMVHRIDSDTFICHTITQSFAVGSISPRDFIDLVYIKRYEGNMNIISSKSVDFPEYPPSSNYIRGYNHPCGFVCSPMEENPAYSKLVMFVQTEMRGKLSPSIIEKTMPSNLVNFILNAKDGIKAHRTPSRRGFHHNSHS | |
RUO | |
STARD6 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
147323 | |
STARD6 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
STARD6 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |