Learn More
Abnova™ Human SLC26A3 Partial ORF (NP_000102, 503 a.a. - 600 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00001811-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene is a transmembrane glycoprotein that transports chloride ions across the cell membrane in exchange for bicarbonate ions. It is localized to the mucosa of the lower intestinal tract, particularly to the apical membrane of columnar epithelium and some goblet cells. The protein is essential for intestinal chloride absorption, and mutations in this gene have been associated with congenital chloride diarrhea. [provided by RefSeq]
Sequence: TQFPKCSTLANIGRTNIYKNKKDYYDMYEPEGVKIFRCPSPIYFANIGFFRRKLIDAVGFSPLRILRKRNKALRKIRKLQKQGLLQVTPKGFICTVDTSpecifications
NP_000102 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.52kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
TQFPKCSTLANIGRTNIYKNKKDYYDMYEPEGVKIFRCPSPIYFANIGFFRRKLIDAVGFSPLRILRKRNKALRKIRKLQKQGLLQVTPKGFICTVDT | |
RUO | |
SLC26A3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
1811 | |
SLC26A3 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CLD/DRA | |
SLC26A3 | |
Recombinant | |
wheat germ expression system |