missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human SLC26A3 Partial ORF (NP_000102, 503 a.a. - 600 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4115.00 NOK - 6245.00 NOK
Specifications
Accession Number | NP_000102 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 1811 |
Molecular Weight (g/mol) | 36.52kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16124181
|
Abnova™
H00001811-Q01.25UG |
25 ug |
6245.00 NOK
25µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16114181
|
Abnova™
H00001811-Q01.10UG |
10 ug |
4115.00 NOK
10µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The protein encoded by this gene is a transmembrane glycoprotein that transports chloride ions across the cell membrane in exchange for bicarbonate ions. It is localized to the mucosa of the lower intestinal tract, particularly to the apical membrane of columnar epithelium and some goblet cells. The protein is essential for intestinal chloride absorption, and mutations in this gene have been associated with congenital chloride diarrhea. [provided by RefSeq]
Sequence: TQFPKCSTLANIGRTNIYKNKKDYYDMYEPEGVKIFRCPSPIYFANIGFFRRKLIDAVGFSPLRILRKRNKALRKIRKLQKQGLLQVTPKGFICTVDTSpecifications
NP_000102 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.52kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CLD/DRA | |
SLC26A3 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
1811 | |
SLC26A3 (Human) Recombinant Protein (Q01) | |
TQFPKCSTLANIGRTNIYKNKKDYYDMYEPEGVKIFRCPSPIYFANIGFFRRKLIDAVGFSPLRILRKRNKALRKIRKLQKQGLLQVTPKGFICTVDT | |
RUO | |
SLC26A3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |