missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human Serpin A3/Alpha-1-Antichymotrypsin ELISA Kit
Description
Human Serpin A3/Alpha-1-Antichymotrypsin quantitates human Serpin A3/Alpha-1-Antichymotrypsin in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Serpin A3/Alpha-1-Antichymotrypsin.
SERPINA encodes Alpha 1-ACT. It is an early-stage acute-phase plasma protein and a serpin that preferentially inactivates chymotrypsin, cathepsin G, and chymase. Alpha-1-ACT, a serine protease inhibitor, is tightly associated with amyloid plaques in Alzheimer's disease (AD) and in normal aged human and monkey brain. Regulation of the serine proteases and serine protease inhibitors plays an important role in neuromuscular differentiation. Prostate specific antigen (PSA), a chymotrypsin-like serine protease, is predominantly complexed to Alpha-1-ACT.
Specifications
Specifications
| Accession Number | AACT; ACT; Alpha-1-antichymotrypsin; Alpha-1-antichymotrypsin His-Pro-less; Cell growth-inhibiting gene 24/25 protein; GIG24; GIG25; growth-inhibiting protein 24; growth-inhibiting protein 25; MGC88254; serine (or cysteine) proteinase inhibitor, clade A, member 3; serpin A3; serpin family A member 3; serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 3; SERPINA3 |
| Assay Range | 0.137 to 100 ng/mL |
| Assay Sensitivity | 0.14 ng/mL |
| Conjugate | HRP |
| Product Type | ELISA |
| Sample Type | Plasma, Serum, Supernatant |
| For Use With (Equipment) | Colorimetric Microplate Reader |
| Gene ID (Entrez) | 12 |
| Gene Symbol | SERPINA3 |
| Interassay CV | <12% |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?