Learn More
Abnova™ Human RCBTB1 Full-length ORF (AAH38104.1, 1 a.a. - 355 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00055213-P01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a protein with an N-terminal RCC1 domain and a C-terminal BTB (broad complex, tramtrack and bric-a-brac) domain. In rat, over-expression of this gene in vascular smooth muscle cells induced cellular hypertrophy. In rat, the C-terminus of RCBTB1 interacts with the angiotensin II receptor-1A. In humans, this gene maps to a region of chromosome 13q that is frequently deleted in B-cell chronic lymphocytic leukemia and other lymphoid malignancies. [provided by RefSeq]
Sequence: MVDVGKWPIFTLLSPQEIASIRKVCVFGTSASEALYVTDNDEVFVFGLNYSNCLGTGDNQSTLVPKKLEGLCGKKIKSLSYGSGPHVLLSTEDGVVYAWGHNGYSQLGNGTTNQGIATVQVCTNLLIKQVVEVACGSHHSMALAADGEVFAWGYNNCGQVGSGSTANQPTPRKVTNCLHIKRVVGIACGQTSSMAVLDNGEVYGWGYNGNGQLGLGNNGNQLTPVRVAALHSVCVNQIVCGYAHTLALTDEGLLYAWGANTYGQLGTGNKNNLLSPAHIMVEKERVVEIAACHSAHTSAAKTQGGHVYMWGQCRGQSVILPHLTHFSCTDDVFACFATPAVSWRLLSVVSIVIGKSpecifications
AAH38104.1 | |
Liquid | |
55213 | |
RCBTB1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MVDVGKWPIFTLLSPQEIASIRKVCVFGTSASEALYVTDNDEVFVFGLNYSNCLGTGDNQSTLVPKKLEGLCGKKIKSLSYGSGPHVLLSTEDGVVYAWGHNGYSQLGNGTTNQGIATVQVCTNLLIKQVVEVACGSHHSMALAADGEVFAWGYNNCGQVGSGSTANQPTPRKVTNCLHIKRVVGIACGQTSSMAVLDNGEVYGWGYNGNGQLGLGNNGNQLTPVRVAALHSVCVNQIVCGYAHTLALTDEGLLYAWGANTYGQLGTGNKNNLLSPAHIMVEKERVVEIAACHSAHTSAAKTQGGHVYMWGQCRGQSVILPHLTHFSCTDDVFACFATPAVSWRLLSVVSIVIGK | |
RUO | |
RCBTB1 | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
64kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CLLD7/CLLL7/GLP/MGC33184/RP11-185C18.1 | |
RCBTB1 | |
Yes | |
wheat germ expression system |